Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Mimosine 98+%

Supplier: AMBEED, INC
Warning

A858029-100MG A858029-1G A858029-250MG A858029-50MG
76807-702EA 154 USD InStock
76807-702 76807-704 76807-706 76807-708
L-Mimosine 98+%
L-Mimosine
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O₄
MW: 198.2 g/mol
MDL Number: MFCD00075909
CAS Number: 500-44-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty