Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Mercaptoethyl)pyrazine 98%

Supplier: AMBEED, INC
Danger

A858887-25G A858887-5G
76807-710EA 84 USD InStock
76807-710 76807-712
(2-Mercaptoethyl)pyrazine 98%
(2-Mercaptoethyl)pyrazine
Precaución: for Research Use Only


Formula: C₆H₈N₂S
MDL Number: MFCD00053160
CAS Number: 35250-53-4
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty