Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Oxo-3,4-dihydrophthalazin-1-yl)acetic acid 95%

Supplier: AMBEED, INC
Warning

A866021-100MG A866021-1G A866021-250MG A866021-5G
76807-746EA 30 USD InStock
76807-746 76807-748 76807-750 76807-752
(4-Oxo-3,4-dihydrophthalazin-1-yl)acetic acid 95%
(4-Oxo-3,4-dihydrophthalazin-1-yl)acetic acid
Precaución: for Research Use Only


Formula: C₁₀H₈N₂O₃
MDL Number: MFCD00085017
CAS Number: 25947-11-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty