Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Bromomethyl)isoquinoline hydrobromide 95%

Supplier: AMBEED, INC
Danger

A881242-100MG A881242-1G A881242-250MG
76807-854EA 43 USD InStock
76807-854 76807-856 76807-858
1-(Bromomethyl)isoquinoline hydrobromide 95%
1-(Bromomethyl)isoquinoline hydrobromide
Precaución: for Research Use Only


Formula: C₁₀H₈BrN·BrH
MDL Number: MFCD03086096
CAS Number: 337508-56-2
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty