Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-d-trp-opfp 98%

Supplier: AMBEED, INC
Warning

A882689-1G A882689-25G A882689-5G
76807-860EA 73.21 USD InStock
76807-860 76807-862 76807-864 76807-866
Fmoc-d-trp-opfp 98%
Fmoc-d-trp-opfp
Precaución: for Research Use Only


Formula: C₃₂H₂₁F₅N₂O₄
MW: 592.522 g/mol
MDL Number: MFCD00155634
CAS Number: 136554-94-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty