Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)benzoyl acetonitrile 95%

Supplier: AMBEED, INC
Danger

A898874-100MG A898874-1G A898874-250MG
76807-918EA 36 USD InStock
76807-918 76807-920 76807-922
4-(Trifluoromethoxy)benzoyl acetonitrile 95%
4-(Trifluoromethoxy)benzoyl acetonitrile
Precaución: for Research Use Only


Formula: C₁₀H₆F₃NO₂
MW: 229.16 g/mol
MDL Number: MFCD00052146
CAS Number: 122454-46-0
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty