Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl phosphate 95%

Supplier: AMBEED, INC
Danger

A919746-100MG A919746-10G A919746-1G A919746-250MG A919746-25G A919746-5G
76808-004EA 7 USD InStock
76808-004 76808-006 76808-008 76808-010 76808-012 76808-014
Diethyl phosphate 95%
Diethyl phosphate
Precaución: for Research Use Only


Formula: C₄H₁₁O₄P
MDL Number: MFCD00039889
CAS Number: 598-02-7
UN: 3265
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty