Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl-2,2'-thiodiglycolate 98%

Supplier: AMBEED, INC
Warning

A926756-1G A926756-25G A926756-5G
76808-028EA 8 USD InStock
76808-028 76808-030 76808-032
Diethyl-2,2'-thiodiglycolate 98%
Diethyl-2,2'-thiodiglycolate
Precaución: for Research Use Only


Formula: C₈H₁₄O₄S
MDL Number: MFCD00087932
CAS Number: 925-47-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty