Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Glyceraldehyde 90%

Supplier: AMBEED, INC
Warning

A951393-1G A951393-250MG A951393-100MG
76808-084EA 116 USD InStock
76808-084 76808-086 77627-774
DL-Glyceraldehyde 90%
DL-Glyceraldehyde
Precaución: for Research Use Only


Formula: C₃H₆O₃
MW: 90.078 g/mol
MDL Number: MFCD00064379
CAS Number: 56-82-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty