Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimesna 98%

Supplier: AMBEED, INC
Warning

A951964-100MG A951964-1MG A951964-25MG
76808-100EA 17 USD InStock
76808-100 76808-102 76808-104
Dimesna 98%
Dimesna
Precaución: for Research Use Only


Formula: C₄H₈O₆S₄·2Na
MW: 326.342 g/mol
MDL Number: MFCD00868218
CAS Number: 16208-51-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty