Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-Cytisine 95%

Supplier: AMBEED, INC
Danger

A969984-100MG A969984-10G A969984-1G A969984-50MG A969984-5G A969984-250MG
76808-146EA 6 USD InStock
76808-146 76808-148 76808-150 76808-152 76808-154 77670-410
(-)-Cytisine 95%
(-)-Cytisine
Precaución: for Research Use Only


Formula: C₁₁H₁₄N₂O
MW: 190.25 g/mol
MDL Number: MFCD00136048
CAS Number: 485-35-8
UN: 1544
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty