Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-1-Pentenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A971597-100MG A971597-1G A971597-250MG A971597-5G
76808-164EA 15 USD InStock
76808-164 76808-166 76808-168 76808-170
(E)-1-Pentenylboronic acid pinacol ester 98%
(E)-1-Pentenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₁H₂₁BO₂
MDL Number: MFCD05663882
CAS Number: 161395-96-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty