Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(((benzyloxy)carbonyl)amino)propanoic acid 98%

Supplier: AMBEED, INC
Warning

A995330-1G A995330-250MG A995330-5G
76808-208EA 51 USD InStock
76808-208 76808-210 76808-212
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(((benzyloxy)carbonyl)amino)propanoic acid 98%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(((benzyloxy)carbonyl)amino)propanoic acid
Precaución: for Research Use Only


Formula: C₂₆H₂₄N₂O₆
MW: 460.47 g/mol
MDL Number: MFCD02684373
CAS Number: 387824-80-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty