Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hemin chloride 97%

Supplier: AMBEED, INC
Warning

A124131-100G A124131-1G A124131-250MG A124131-5G A124131-10G A124131-500G
76797-578EA 715 USD InStock
76797-578 76797-580 76797-582 76797-584 77566-374 77666-770
Hemin chloride 97%
Hemin chloride
Precaución: For research use only.


Formula: C₃₄H₃₂ClFeN₄O₄
MDL Number: MFCD00010726
CAS Number: 16009-13-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty