Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4,6-dimethylnicotinic acid 97%

Supplier: AMBEED, INC
Warning

A102446-10G A102446-1G A102446-250MG A102446-25G A102446-5G
76809-324EA 174 USD InStock
76809-324 76809-326 76809-328 76809-330 76809-332
2-Chloro-4,6-dimethylnicotinic acid 97%
2-Chloro-4,6-dimethylnicotinic acid
Precaución: for Research Use Only


Formula: C₈H₈ClNO₂
MDL Number: MFCD02110082
CAS Number: 66662-48-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty