Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3H-[1,2,3]triazolo[4,5-c]pyridine 98%

Supplier: AMBEED, INC
Warning

A102571-1G A102571-250MG
76809-336EA 186 USD InStock
76809-336 76809-338
4-Chloro-3H-[1,2,3]triazolo[4,5-c]pyridine 98%
4-Chloro-3H-[1,2,3]triazolo[4,5-c]pyridine
Precaución: for Research Use Only


Formula: C₅H₃ClN₄
MW: 154.557 g/mol
MDL Number: MFCD13193520
CAS Number: 36258-82-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty