Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromo-6-methoxypyridin-2-yl)methanol 95%

Supplier: AMBEED, INC
Warning

A103765-100MG A103765-10G A103765-1G A103765-250MG A103765-5G
76809-420EA 11 USD InStock
76809-420 76809-422 76809-424 76809-426 76809-428
(3-Bromo-6-methoxypyridin-2-yl)methanol 95%
(3-Bromo-6-methoxypyridin-2-yl)methanol
Precaución: for Research Use Only


Formula: C₇H₈BrNO₂
MDL Number: MFCD12406110
CAS Number: 623942-84-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty