Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Difluoropropanoic acid 95%

Supplier: AMBEED, INC
Danger

A104150-100MG A104150-1G A104150-250MG
76809-470EA 70 USD InStock
76809-470 76809-472 76809-474
3,3-Difluoropropanoic acid 95%
3,3-Difluoropropanoic acid
Precaución: for Research Use Only


Formula: C₃H₄F₂O₂
MDL Number: MFCD12024562
CAS Number: 155142-69-1
UN: 3265
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty