Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methoxycarbonyl)cyclohexanecarboxylic acid 97%

Supplier: AMBEED, INC

A104845-100MG A104845-1G A104845-250MG A104845-5G
76809-524EA 18 USD InStock
76809-524 76809-526 76809-528 76809-530
3-(Methoxycarbonyl)cyclohexanecarboxylic acid 97%
3-(Methoxycarbonyl)cyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₉H₁₄O₄
MDL Number: MFCD17170191
CAS Number: 25090-39-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty