Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol 97%

Supplier: AMBEED, INC
Warning

A105335-100G A105335-1MG
76809-550EA 195 USD InStock
76809-550 76809-552 76809-554
(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol 97%
(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
Precaución: for Research Use Only


Formula: HOH₂C(CH(OH))₄CH₂OH
MDL Number: MFCD00064288
CAS Number: 2234851-14-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty