Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrazolo[1,5-a]pyrimidin-3-amine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A105394-100MG A105394-1G A105394-250MG
76809-560EA 29 USD InStock
76809-560 76809-562 76809-564
Pyrazolo[1,5-a]pyrimidin-3-amine hydrochloride 95%
Pyrazolo[1,5-a]pyrimidin-3-amine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₇ClN₄
MDL Number: MFCD21604217
CAS Number: 232600-78-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty