Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,4-dioxo-4-(o-tolyl)butanoate 95%

Supplier: AMBEED, INC
Warning

A105461-100MG A105461-1G A105461-250MG A105461-5G
76809-566EA 37 USD InStock
76809-566 76809-568 76809-570 76809-572
Methyl 2,4-dioxo-4-(o-tolyl)butanoate 95%
Methyl 2,4-dioxo-4-(o-tolyl)butanoate
Precaución: for Research Use Only


Formula: C₁₂H₁₂O₄
MDL Number: MFCD11188866
CAS Number: 1037130-77-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty