Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl thiazol-4-ylcarbamate 98%

Supplier: AMBEED, INC
Warning

A105698-100G A105698-1G A105698-250MG A105698-25G A105698-5G
76809-602EA 387 USD InStock
76809-602 76809-604 76809-606 76809-608 76809-610
tert-Butyl thiazol-4-ylcarbamate 98%
tert-Butyl thiazol-4-ylcarbamate
Precaución: for Research Use Only


Formula: C₈H₁₂N₂O₂S
MW: 200.2600 g/mol
MDL Number: MFCD18800853
CAS Number: 1235406-42-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty