Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl [2,2'-bipyridine]-4,4'-dicarboxylate 98%

Supplier: AMBEED, INC
Warning

A105731-1G A105731-250MG A105731-5G A105731-100G
76809-612EA 9 USD InStock
76809-612 76809-614 76809-616 77670-518
Dimethyl [2,2'-bipyridine]-4,4'-dicarboxylate 98%
Dimethyl [2,2'-bipyridine]-4,4'-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₄H₁₂N₂O₄
MW: 272.2600 g/mol
MDL Number: MFCD02669916
CAS Number: 71071-46-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty