Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Bis(9H-carbazol-9-yl)biphenyl 97%

Supplier: AMBEED, INC
Warning

A105823-10G A105823-1G A105823-25G A105823-5G
76809-618EA 49 USD InStock
76809-618 76809-620 76809-622 76809-624
4,4'-Bis(9H-carbazol-9-yl)biphenyl 97%
4,4'-Bis(9H-carbazol-9-yl)biphenyl
Precaución: for Research Use Only


Formula: C₃₆H₂₄N₂
MDL Number: MFCD00093417
CAS Number: 58328-31-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty