Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Z)-2-(2-Aminothiazol-4-yl)-2-(hydroxyimino)acetic acid 95%

Supplier: AMBEED, INC
Danger

A107950-1G A107950-25G A107950-5G
76809-780EA 55.17 USD InStock
76809-780 76809-782 76809-784 76809-786
(Z)-2-(2-Aminothiazol-4-yl)-2-(hydroxyimino)acetic acid 95%
(Z)-2-(2-Aminothiazol-4-yl)-2-(hydroxyimino)acetic acid
Precaución: for Research Use Only


Formula: C₅H₅N₃O₃S
MW: 187.18 g/mol
MDL Number: MFCD00062754
CAS Number: 66338-96-3
UN: 2926
ADR: 4.1,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty