Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium (R)-2-(6-(4-chlorophenoxy)hexyl)oxirane-2-carboxylate 98%

Supplier: AMBEED, INC
Warning

A108121-10MG A108121-1MG A108121-50MG A108121-5MG
76809-800EA 106 USD InStock
76809-800 76809-802 76809-804 76809-806
Sodium (R)-2-(6-(4-chlorophenoxy)hexyl)oxirane-2-carboxylate 98%
Sodium (R)-2-(6-(4-chlorophenoxy)hexyl)oxirane-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₅H₁₈ClNaO₄
MDL Number: MFCD07787411
CAS Number: 828934-41-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty