Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dihydroxy-5-nitropyrimidine 98%

Supplier: AMBEED, INC
Warning

A109052-100G A109052-10G A109052-25G A109052-5G
76809-836EA 94 USD InStock
76809-836 76809-838 76809-840 76809-842
4,6-Dihydroxy-5-nitropyrimidine 98%
4,6-Dihydroxy-5-nitropyrimidine
Precaución: for Research Use Only


Formula: C₄H₃N₃O₄
MW: 157.09 g/mol
Melting Pt: 300
MDL Number: MFCD00006110
CAS Number: 2164-83-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty