Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzyloxy)-2-fluorobenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A110819-10G A110819-1G A110819-250MG
76809-916EA 433 USD InStock
76809-916 76809-918 76809-920
4-(Benzyloxy)-2-fluorobenzaldehyde 98%
4-(Benzyloxy)-2-fluorobenzaldehyde
Precaución: for Research Use Only


Formula: C₁₄H₁₁FO₂
MW: 230.23 g/mol
MDL Number: MFCD12025062
CAS Number: 504414-32-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty