Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(tert-Butoxycarbonyl)-1,4-diaminobutane 97% (by GC)

Supplier: AMBEED, INC
Danger

A111705-10G A111705-1G A111705-5G
76809-952EA 24 USD InStock
76809-952 76809-954 76809-956
N-(tert-Butoxycarbonyl)-1,4-diaminobutane 97% (by GC)
N-(tert-Butoxycarbonyl)-1,4-diaminobutane
Precaución: for Research Use Only


Formula: C₉H₂₀N₂O₂
MDL Number: MFCD00210019
CAS Number: 68076-36-8
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty