Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-(1-Benzylazetidine-2,4-diyl)dimethanol 95%

Supplier: AMBEED, INC
Warning

A115209-100MG A115209-1G A115209-250MG A115209-5G
76810-112EA 7 USD InStock
76810-112 76810-114 76810-116 76810-118
cis-(1-Benzylazetidine-2,4-diyl)dimethanol 95%
cis-(1-Benzylazetidine-2,4-diyl)dimethanol
Precaución: for Research Use Only


Formula: C₁₂H₁₇NO₂
MDL Number: MFCD28991841
CAS Number: 127310-66-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty