Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,3R,4S,5R)-3,4-Bis((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,5-dihydroxycyclohexanecarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A117820-1MG A117820-5MG
76810-280EA 22 USD InStock
76810-280 76810-282
(1R,3R,4S,5R)-3,4-Bis((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,5-dihydroxycyclohexanecarboxylic acid 98%
(1R,3R,4S,5R)-3,4-Bis((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,5-dihydroxycyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₂₅H₂₄O₁₂
MDL Number: MFCD10566623
CAS Number: 57378-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty