Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromo-1-hydroxynaphthalene 97%

Supplier: AMBEED, INC
Warning

A119332-100MG A119332-1G A119332-250MG
76810-374EA 47 USD InStock
76810-374 76810-376 76810-378
7-Bromo-1-hydroxynaphthalene 97%
7-Bromo-1-hydroxynaphthalene
Precaución: for Research Use Only


Formula: C₁₀H₇BrO
MDL Number: MFCD17012439
CAS Number: 91270-69-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty