Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Bromomethyl)benzo[d][1,3]dioxole 95%

Supplier: AMBEED, INC
Danger

A125880-250MG A125880-5G
76810-906EA 75 USD InStock
76810-906 76810-908
4-(Bromomethyl)benzo[d][1,3]dioxole 95%
4-(Bromomethyl)benzo[d][1,3]dioxole
Precaución: for Research Use Only


Formula: C₈H₇BrO₂
MW: 215.04 g/mol
MDL Number: MFCD21604251
CAS Number: 101417-40-7
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty