Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-1,3-azetidinedicarboxylic acid tert-butyl ester 98%

Supplier: AMBEED, INC
Warning

A127335-100MG A127335-1G A127335-250MG
76810-992EA 27 USD InStock
76810-992 76810-994 76810-996
3-Fluoro-1,3-azetidinedicarboxylic acid tert-butyl ester 98%
3-Fluoro-1,3-azetidinedicarboxylic acid tert-butyl ester
Precaución: for Research Use Only


Formula: C₉H₁₄FNO₄
MDL Number: MFCD15071784
CAS Number: 1126650-67-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty