Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-oxocyclobutanecarboxylate 97%

Supplier: AMBEED, INC
Warning

A127601-100G A127601-10G A127601-1G A127601-25G A127601-5G
76810-998EA 107 USD InStock
76810-998 76811-000 76811-002 76811-004 76811-006
Ethyl-3-oxocyclobutanecarboxylate 97%
Ethyl-3-oxocyclobutanecarboxylate
Precaución: for Research Use Only


Formula: C₇H₁₀O₃
MDL Number: MFCD09834114
CAS Number: 87121-89-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty