Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1,N1,N2-Trimethylethane-1,2-diamine dihydrochloride 97%

Supplier: AMBEED, INC
Warning

A128313-10G A128313-1G A128313-5G
76811-048EA 130 USD InStock
76811-048 76811-050 76811-052 76811-054
N1,N1,N2-Trimethylethane-1,2-diamine dihydrochloride 97%
N1,N1,N2-Trimethylethane-1,2-diamine dihydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₆Cl₂N₂
MDL Number: MFCD21604009
CAS Number: 326888-32-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty