Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-iodobenzonitrile 97%

Supplier: AMBEED, INC
Warning

A128384-10G A128384-1G A128384-250MG A128384-5G
76811-056EA 5.31 USD InStock
76811-056 76811-058 76811-060 76811-062 76811-064
4-Chloro-3-iodobenzonitrile 97%
4-Chloro-3-iodobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃ClIN
MDL Number: MFCD09025674
CAS Number: 914106-26-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty