Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2',6'-Dimethoxy-[1,1'-biphenyl]-2-yl)diphenylphosphine 97%

Supplier: AMBEED, INC
Warning

A129382-100MG A129382-1G A129382-250MG A129382-5G A129382-25G
76811-100EA 5 USD InStock
76811-100 76811-102 76811-104 76811-106 77670-830
(2',6'-Dimethoxy-[1,1'-biphenyl]-2-yl)diphenylphosphine 97%
(2',6'-Dimethoxy-[1,1'-biphenyl]-2-yl)diphenylphosphine
Precaución: for Research Use Only


Formula: C₂₆H₂₃O₂P
MDL Number: MFCD22199795
CAS Number: 819867-24-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty