Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-bromo-5-fluorophenol 98%

Supplier: AMBEED, INC
Warning

A130852-1G A130852-100MG
76811-176EA 138 USD InStock
76811-176 77670-848
2-Amino-4-bromo-5-fluorophenol 98%
2-Amino-4-bromo-5-fluorophenol
Precaución: for Research Use Only


Formula: C₆H₅BrFNO
MDL Number: MFCD11101429
CAS Number: 1016234-89-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty