Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzyloxy)-4-bromo-1-fluorobenzene 97%

Supplier: AMBEED, INC
Warning

A131591-100MG A131591-1G A131591-250MG A131591-5G
76811-210EA 13 USD InStock
76811-210 76811-212 76811-214 76811-216
2-(Benzyloxy)-4-bromo-1-fluorobenzene 97%
2-(Benzyloxy)-4-bromo-1-fluorobenzene
Precaución: for Research Use Only


Formula: C₁₃H₁₀BrFO
MW: 281.12 g/mol
MDL Number: MFCD11520671
CAS Number: 1036724-54-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty