Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4-Dimethyl-1,3-cyclohexanedione 97%

Supplier: AMBEED, INC
Warning

A131917-100G A131917-1G A131917-25G
76811-226EA 462 USD InStock
76811-226 76811-228 76811-230
4,4-Dimethyl-1,3-cyclohexanedione 97%
4,4-Dimethyl-1,3-cyclohexanedione
Precaución: for Research Use Only


Formula: C₈H₁₂O₂
MDL Number: MFCD00075224
CAS Number: 562-46-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty