Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chloro-6-methylpyridine 95%

Supplier: AMBEED, INC
Warning

A132888-250MG A132888-1G
76811-310EA 38 USD InStock
76811-310 77670-884
4-Bromo-2-chloro-6-methylpyridine 95%
4-Bromo-2-chloro-6-methylpyridine
Precaución: For research use only.


Formula: C₆H₅BrClN
MW: 206.46 g/mol
MDL Number: MFCD14582031
CAS Number: 1206250-53-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty