Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nε-acetyl-Nα-boc-L-lysine 97%

Supplier: AMBEED, INC
Warning

A132987-10G A132987-1G A132987-250MG A132987-25G
76811-314EA 80 USD InStock
76811-314 76811-316 76811-318 76811-320
Nε-acetyl-Nα-boc-L-lysine 97%
Nepsilon-acetyl-Nalpha-boc-L-lysine
Precaución: for Research Use Only


Formula: C₁₃H₂₄N₂O₅
MW: 288.344 g/mol
MDL Number: MFCD00057791
CAS Number: 6404-26-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty