Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,6-Dihydroxynaphthalene 97%

Supplier: AMBEED, INC
Warning

A134551-100G A134551-10G A134551-25G
76811-418EA 36 USD InStock
76811-418 76811-420 76811-422
1,6-Dihydroxynaphthalene 97%
1,6-Dihydroxynaphthalene
Precaución: for Research Use Only


Formula: C₁₀H₈O₂
MDL Number: MFCD00003981
CAS Number: 575-44-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty