Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Methyl 1-Tritylaziridine-2-carboxylate 97%

Supplier: AMBEED, INC
Warning

A134664-100G A134664-10G A134664-1G A134664-250MG A134664-25G A134664-5G
76811-428EA 267 USD InStock
76811-428 76811-430 76811-432 76811-434 76811-436 76811-438
(S)-Methyl 1-Tritylaziridine-2-carboxylate 97%
(S)-Methyl 1-Tritylaziridine-2-carboxylate
Precaución: for Research Use Only


Formula: C₂₃H₂₁NO₂
MDL Number: MFCD01863736
CAS Number: 75154-68-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty