Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,7-Dichlorothieno[2,3-d]pyridazine 95%

Supplier: AMBEED, INC
Warning

A136387-100MG A136387-1G A136387-250MG A136387-5G
76811-540EA 18 USD InStock
76811-540 76811-542 76811-544 76811-546
4,7-Dichlorothieno[2,3-d]pyridazine 95%
4,7-Dichlorothieno[2,3-d]pyridazine
Precaución: for Research Use Only


Formula: C₆H₂Cl₂N₂S
MDL Number: MFCD09965522
CAS Number: 699-89-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty