Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-4-chloropicolinic acid 97%

Supplier: AMBEED, INC
Warning

A139259-100MG A139259-1G A139259-50MG
76811-684EA 22 USD InStock
76811-684 76811-686 76811-688
5-Bromo-4-chloropicolinic acid 97%
5-Bromo-4-chloropicolinic acid
Precaución: for Research Use Only


Formula: C₆H₃BrClNO₂
MDL Number: MFCD13189053
CAS Number: 1060802-25-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty