Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromocytosine 95%

Supplier: AMBEED, INC
Danger

A140809-100MG A140809-10G A140809-1G A140809-250MG A140809-25G A140809-5G
76811-762EA 5 USD InStock
76811-762 76811-764 76811-766 76811-768 76811-770 76811-772
5-Bromocytosine 95%
5-Bromocytosine
Precaución: for Research Use Only


Formula: C₄H₄BrN₃O
MDL Number: MFCD00056025
CAS Number: 2240-25-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty