Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Benzyl-3-hydroxy-1-(methylamino)-1-oxopropan-2-ylcarbamate 95%

Supplier: AMBEED, INC
Warning

A144699-1G A144699-250MG A144699-5G
76811-916EA 94 USD InStock
76811-916 76811-918 76811-920
(S)-Benzyl-3-hydroxy-1-(methylamino)-1-oxopropan-2-ylcarbamate 95%
(S)-Benzyl-3-hydroxy-1-(methylamino)-1-oxopropan-2-ylcarbamate
Precaución: for Research Use Only


Formula: C₁₂H₁₆N₂O₄
MW: 252.27 g/mol
MDL Number: MFCD17011849
CAS Number: 19647-68-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty